Learn More
BINDER™ Arabidopsis Illumination
Arabidopsis Illumination - Illumination cassettes equipped with Arabidopsis fluorescent tubes instead of daylight fluorescent tubes
Brand: BINDER™ 8012-0686
Additional Details : Weight : 0.50000kg
Description
- 17 W or 25 W option model
- For use with a range of growth chambers
Specifications
Arabidopsis illumination - Illumination cassettes equipped with Arabidopsis fluorescent tubes instead of daylight fluorescent tubes | |
BINDER Chambers KBW 720 | |
Fluorescent Light | |
25 W |