Learn More
Hirschmann™ Class AS Graduated Pipettes with Amber Graduation
Provides volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Class AS Graduated Pipettes with Amber Graduation come with a cotton plug end as standard.
POA
Product Code | Brand | Volume (Metric) | Graduations | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Graduations | Price | Quantity & Availability | ||||
11766632
|
Hirschmann™
1100229 |
0.5 mL | 0.01 mL |
POA
Pack of 12 |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
11792262
|
Hirschmann™
1100233 |
1 mL | 0.01 mL |
POA
Pack of 12 |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
11722272
|
Hirschmann™
1100260 |
10 mL | 0.1 mL |
POA
Pack of 12 |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
11732272
|
Hirschmann™
1100270 |
20 mL | 0.1 mL |
POA
Pack of 6 |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
11796632
|
Hirschmann™
1100254 |
5 mL | 0.1 mL |
POA
Pack of 12 |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
11702272
|
Hirschmann™
1100241 |
2 mL | 0.02 mL |
POA
Pack of 12 |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
11712272
|
Hirschmann™
1100253 |
5 mL | 0.05 mL |
POA
Pack of 12 |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
11776632
|
Hirschmann™
1100235 |
1 mL | 0.1 mL |
POA
Pack of 12 |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
11786632
|
Hirschmann™
1100242 |
2 mL | 0.1 mL |
POA
Pack of 12 |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | |||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
Description
- Main point ring graduation
- Graduated not to tip, zero on top
- Acc. to DIN EN ISO 835
- Conformity certified, with dated batch identification
Specifications
AS | |
Yes | |
Amber |