Learn More
VACUUBRAND™ Aluminum Clamping Rings
A rapid connection that can be mounted without any tools for soft seals and indium seals.
21.55 EUR - 25.45 EUR
Specifications
For Use With | For Small KF Flange |
---|---|
Material | Aluminum |
Quantity | 1 Each |
Type | Clamping Ring |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
10518381
|
VACUUBRAND™
20660001 |
DN 20/25 mm |
25.45 EUR
Each |
Estimated Shipment: 05-11-2024
Log in to see stock. |
|||||
10419122
|
VACUUBRAND™
20660000 |
DN 10/16 mm |
21.55 EUR
Each |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
Description
- Provided with wing nut
- Available in two different sizes
Specifications
For Small KF Flange | |
Aluminum | |
1 Each | |
Clamping Ring |