Learn More
Al-23 Tube, Both ends open, O.D.: 60mm, I.D.: 50mm, Thermo Scientific™
98.10 EUR - 952.00 EUR
Specifications
Chemical Name or Material Al-23 Tube
Name Note Both ends open
View More Specs
Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11304057
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031581.DM
100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11374047
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031581.DR
200 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11354047
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031581.DW
300 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11314057
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031581.EH
600 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11394047
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031581.EM
800 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11384047
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031581.EO
1000 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Al-23 Tube
Both ends open
Videos
Safety and Handling
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures