Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Ultrafiltration
Optimized for a variety of applications requiring larger per well volume processing.
422.00 EUR - 515.00 EUR
Specifications
Material (Membrane) | Omega (Modified PES) |
---|---|
Sterility | Non-sterile |
Housing Material | Polypropylene Housing |
Product Code | Brand | Volume (Metric) | MWCO | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | MWCO | Price | Quantity & Availability | ||||
17194601
|
Cytiva (formerly Pall Lab)
8035 |
350 μL | 30 kDa |
515.00 EUR
Pack of 10 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
17184601
|
Cytiva (formerly Pall Lab)
8034 |
350 μL | 10 kDa |
515.00 EUR
Pack of 10 |
Estimated Shipment: 22-07-2024
Log in to see stock. |
||||
17104611
|
Cytiva (formerly Pall Lab)
8036 |
350 μL | 100 kDa |
515.00 EUR
Pack of 10 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
||||
17184621
|
Cytiva (formerly Pall Lab)
8163 |
1 mL | 3 kDa |
441.00 EUR
Pack of 5 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
17174601
|
Cytiva (formerly Pall Lab)
8033 |
350 μL | 3 kDa |
515.00 EUR
Pack of 10 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17104631
|
Cytiva (formerly Pall Lab)
8165 |
1 mL | 30 kDa |
422.00 EUR
Pack of 5 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
||||
17194621
|
Cytiva (formerly Pall Lab)
8164 |
1 mL | 10 kDa |
446.00 EUR
Pack of 5 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
17114631
|
Cytiva (formerly Pall Lab)
8166 |
1 mL | 100 kDa |
422.00 EUR
Pack of 5 |
Estimated Shipment: 09-09-2024
Log in to see stock. |
||||
Description
- Plates are constructed of chemically resistant, biologically-inert polypropylene
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Omega (Modified PES) | |
Non-sterile | |
Polypropylene Housing |