Learn More
Cytiva (formerly Pall Lab) AcroPak™ 200 Capsules with Fluorodyne™ II Membrane
AcroPak™ 200 Capsules designed for research applications only.
Brand:  Cytiva (formerly Pall Lab) 12069
Additional Details : Weight : 0.44650kg
Product Code. 17154211
  • 361.00 EUR / Pack of 3
Estimated Shipment: 17-10-2024
Description
  • Efficient processing volumes up to 20 L
  • Fluorodyne II (PVDF) membrane is compatible with aqueous and many organic solvents
  • AcroPak 20 filters are designed to add convenience to small- and medium-volume filtrations
Specifications
Specifications
Hydrophilic PVDF
Sterile
3/Pk.
0.2 μm
200 cm2
Stepped Hose Barb