Learn More
Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with Versapor™ Membrane
For use in cell culture and other sensitive biological research.
254.00 EUR - 2875.00 EUR
Specifications
Material (Membrane) | Acrylic Copolymer (Hydrophilic) |
---|---|
Sterility | Non-sterile |
Product Code | Brand | Pore Size | Diameter (Metric) | No. per Pack | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Diameter (Metric) | No. per Pack | Price | Quantity & Availability | |||
17194321
|
Cytiva (formerly Pall Lab)
4487 |
0.45 μm | 25 mm | 75/Pk. |
254.00 EUR
Pack of 75 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||
17194311
|
Cytiva (formerly Pall Lab)
4459 |
0.8 μm | 13 mm | 100/Pk. |
675.00 EUR
Pack of 100 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||
17114331
|
Cytiva (formerly Pall Lab)
4489 |
5 μm | 25 mm | 75/Pk. |
254.00 EUR
Pack of 75 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||
17154271
|
Cytiva (formerly Pall Lab)
4189 |
0.8 μm | 25 mm | 75/Pk. |
254.00 EUR
Pack of 75 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||
17124361
|
Cytiva (formerly Pall Lab)
4568 |
0.8 μm | 25 mm | 1000/Pk. |
2875.00 EUR
Pack of 1000 |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||
17134321
|
Cytiva (formerly Pall Lab)
4473 |
0.45 μm | 4 mm | 1000/Pk. |
635.00 EUR
Pack of 250 |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||
17104331
|
Cytiva (formerly Pall Lab)
4488 |
1.2 μm | 25 mm | 75/Pk. |
254.00 EUR
Pack of 75 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||
Description
- Meets aqueous sample filtration needs
- Designed for laboratory applications only
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Acrylic Copolymer (Hydrophilic) | |
Non-sterile |