Learn More
Cytiva (formerly Pall Lab) Acrodisc™ Syringe Filters with Ultipor Membrane Optimized for Scale Up
Syringe filter with Ultipor™ VF Grade DV20 membrane validated for virus removal.
Brand: Cytiva (formerly Pall Lab) 4906
Additional Details : Weight : 0.30000kg
Description
- Suitable for small-scale membrane qualification studies such as bacteriophage or prion (TSE agent) clearance, as well as membrane flow rate, capacity and protein transmission studies
Specifications
0.2 μm | |
Sterile |