Learn More
5-Bromopyridine-3-boronic acid, tech. 85%, Thermo Scientific Chemicals
52.10 EUR - 528.00 EUR
Chemical Identifiers
CAS | 452972-09-7 |
---|---|
Molecular Formula | C5H5BBrNO2 |
Molecular Weight (g/mol) | 201.81 |
MDL Number | MFCD02685634 |
InChI Key | ICCGFOKNFZWCTJ-UHFFFAOYSA-N |
Synonym | 5-bromopyridine-3-boronic acid, 5-bromopyridin-3-yl boronic acid, 3-bromopyridine-5-boronic acid, 3-bromo-5-pyridineboronic acid, 3-bromopyridyl-5-boronic acid, 5-bromo-3-pyridyl boronic acid, 3-bromo-5-pyridylboronic acid, boronic acid, 5-bromo-3-pyridinyl, 5-bromopyridine-3-boronicacid, pubchem5305 |
PubChem CID | 3814811 |
IUPAC Name | (5-bromopyridin-3-yl)boronic acid |
SMILES | OB(O)C1=CN=CC(Br)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11374516
|
Thermo Scientific Alfa Aesar
L20084.MD |
250 mg | |||||||
11354516
|
Thermo Scientific Alfa Aesar
L20084.03 |
1 g |
152.00 EUR
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11364516
|
Thermo Scientific Alfa Aesar
L20084.06 |
5 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
452972-09-7 | |
201.81 | |
ICCGFOKNFZWCTJ-UHFFFAOYSA-N | |
3814811 | |
OB(O)C1=CN=CC(Br)=C1 |
C5H5BBrNO2 | |
MFCD02685634 | |
5-bromopyridine-3-boronic acid, 5-bromopyridin-3-yl boronic acid, 3-bromopyridine-5-boronic acid, 3-bromo-5-pyridineboronic acid, 3-bromopyridyl-5-boronic acid, 5-bromo-3-pyridyl boronic acid, 3-bromo-5-pyridylboronic acid, boronic acid, 5-bromo-3-pyridinyl, 5-bromopyridine-3-boronicacid, pubchem5305 | |
(5-bromopyridin-3-yl)boronic acid |
Specifications
452972-09-7 | |
C5H5BBrNO2 | |
9119873 | |
ICCGFOKNFZWCTJ-UHFFFAOYSA-N | |
(5-bromopyridin-3-yl)boronic acid | |
3814811 | |
85% | |
5-Bromopyridine-3-boronic acid |
>300°C | |
MFCD02685634 | |
5-bromopyridine-3-boronic acid, 5-bromopyridin-3-yl boronic acid, 3-bromopyridine-5-boronic acid, 3-bromo-5-pyridineboronic acid, 3-bromopyridyl-5-boronic acid, 5-bromo-3-pyridyl boronic acid, 3-bromo-5-pyridylboronic acid, boronic acid, 5-bromo-3-pyridinyl, 5-bromopyridine-3-boronicacid, pubchem5305 | |
OB(O)C1=CN=CC(Br)=C1 | |
201.81 | |
201.82 | |
Technical |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only