missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethoxy)benzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 11430847
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139301-27-2
Molecular Formula C7H6BF3O3
Molecular Weight (g/mol) 205.93
MDL Number MFCD01074648
InChI Key HUOFUOCSQCYFPW-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy phenylboronic acid, 4-trifluoromethoxyphenylboronic acid, 4-trifluoromethoxy benzeneboronic acid, 4-trifluoromethoxy phenyl boronic acid, 4-trifluoromethoxy phenyl boranediol, p-trifluoromethoxy phenylboronic acid, 4-trifluoromethoxy-phenyl boronic acid, boronic acid, 4-trifluoromethoxy phenyl, 4-trifluoromethoxy phenylboronicacid
PubChem CID 2734386
IUPAC Name [4-(trifluoromethoxy)phenyl]boronic acid
SMILES OB(O)C1=CC=C(OC(F)(F)F)C=C1

Specifications

Melting Point ∼120°C to 125°C
Quantity 5 g
Beilstein 8548201
Formula Weight 205.93
Percent Purity 98%
Chemical Name or Material 4-(Trifluoromethoxy)benzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'