missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-3-fluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 11302829
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 843663-18-3
Molecular Formula C7H5BFNO2
Molecular Weight (g/mol) 164.93
MDL Number MFCD03411549
InChI Key DECWLXUOZUMPBF-UHFFFAOYSA-N
Synonym 4-cyano-3-fluorobenzeneboronic acid, 4-cyano-3-fluorophenyl boronic acid, 4-borono-2-fluorobenzonitrile, 4-cyano-3-fluoro-phenyl boronic acid, 4-cyano-3-fluoro phenyl boronic acid, 3-fluoro-4-cyanophenylboronic acid, boronic acid, 4-cyano-3-fluorophenyl, 4-cyano-3-fluorophenylboronicacid, pubchem1801, acmc-209pug
PubChem CID 2783130
IUPAC Name (4-cyano-3-fluorophenyl)boronic acid
SMILES OB(O)C1=CC(F)=C(C=C1)C#N

Specifications

Melting Point >300°C
Quantity 5 g
Formula Weight 164.93
Percent Purity 97%
Chemical Name or Material 4-Cyano-3-fluorobenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'