Learn More
3M™ Disposable Protective Oversleeve 444
Provides basic barrier protection
Brand: 3M™ 7000089717
Additional Details : Weight : 3.66600kg
Description
Helps protect arms against light liquid splashes and hazardous dusts
- Tapered design and elastic around top and bottom of sleeve for secure and easy fit
- Anti-static treated