missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diaminobenzidine, 98+%, Thermo Scientific Chemicals

Catalog Number p-7035793
Click to view available options
Quantity:
5 g
25 g
100 g
91-95-2
C12H14N4
214.272
MFCD00007725
HSTOKWSFWGCZMH-UHFFFAOYSA-N
3,3'-diaminobenzidine, 3,3',4,4'-tetraaminobiphenyl, 3,3'-diaminobenzidene, 3,3',4,4'-diphenyltetramine, 3,3',4,4'-tetraminobiphenyl, 3,3',4,4'-tetraaminodiphenyl, 1,1'-biphenyl-3,3',4,4'-tetramine, biphenyl-3,3',4,4'-tetrayltetraamine, unii-2rv4t6khqi, 4-3,4-diaminophenyl benzene-1,2-diamine
7071
4-(3,4-diaminophenyl)benzene-1,2-diamine
C1=CC(=C(C=C1C2=CC(=C(C=C2)N)N)N)N
91-95-2
C12H14N4
214.272
MFCD00007725
HSTOKWSFWGCZMH-UHFFFAOYSA-N
3,3'-diaminobenzidine, 3,3',4,4'-tetraaminobiphenyl, 3,3'-diaminobenzidene, 3,3',4,4'-diphenyltetramine, 3,3',4,4'-tetraminobiphenyl, 3,3',4,4'-tetraaminodiphenyl, 1,1'-biphenyl-3,3',4,4'-tetramine, biphenyl-3,3',4,4'-tetrayltetraamine, unii-2rv4t6khqi, 4-3,4-diaminophenyl benzene-1,2-diamine
7071
4-(3,4-diaminophenyl)benzene-1,2-diamine
C1=CC(=C(C=C1C2=CC(=C(C=C2)N)N)N)N

3,3'-Diaminobenzidine is used as peroxidase substrate and a reagent for spectrophotometric determination of selenium. DAB is used for the immunohistochemical staining of nucleic acids and proteins. As a dark brown dye, was used as an antibody-specific stain to identify the paired antibodies in breast tissue. It may be used for the synthesis of a useful, linear, Schiff-base coordination polymer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′-Diaminobenzidine is used as peroxidase substrate and a reagent for spectrophotometric determination of selenium. DAB is used for the immunohistochemical staining of nucleic acids and proteins. As a dark brown dye, was used as an antibody-specific stain to identify the paired antibodies in breast tissue. It may be used for the synthesis of a useful, linear, Schiff-base coordination polymer.

Solubility
Soluble in water (partly).

Notes
Light sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY
Melting Point 175°C to 177°C
Quantity 5 g
Sensitivity Light sensitive
Solubility Information Soluble in water (partly).
Formula Weight 214.27
Percent Purity 99%
Chemical Name or Material 3,3'-Diaminobenzidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'