Learn More
Merck Accessories for Glass Microanalysis Filter Holders
Accessories for filter holders for sample preparation.
97.60 EUR - 464.00 EUR
Specifications
Autoclavable | Not Autoclavable |
---|---|
For Use With (Equipment) | 25 mm glass microanalysis vacuum filter |
Sterility | Non-sterile |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
15874146
|
Merck
XX1012503 |
Spring Clamp |
97.60 EUR
Each |
In Stock
Log in to see stock. |
|||||
15804156
|
Merck
XX1012508 |
Perforated Stopper |
192.00 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
15854156
|
Merck
XX1012542 |
Base, Stopper, Screen and Gasket |
464.00 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
15834156
|
Merck
XX1012532 |
Base, Stopper and Screen |
403.00 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
15814156
|
Merck
XX1012514 |
Funnel |
193.00 EUR
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
15864146
|
Merck
XX1012502 |
Base with Stopper |
324.00 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Description
Accessories for filter holders for sample preparation.
Specifications
Not Autoclavable | |
25 mm glass microanalysis vacuum filter | |
Non-sterile |