Learn More
2-Fluoro-4-(trifluoromethyl)nicotinic acid, 97%, Thermo Scientific Chemicals
Product Code. 15465367
Click to view available options
Quantity:
250 mg
1 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1040681-74-0
Molecular Formula C7H3F4NO2
Molecular Weight (g/mol) 209.1
MDL Number MFCD10687403
InChI Key ABXMDMDQUVWALB-UHFFFAOYSA-N
Synonym 2-fluoro-4-trifluoromethyl pyridine-3-carboxylic acid, 2-fluoro-4-trifluoromethyl pyridine-3-carboxylicacid, 2-fluoro-4-trifluoromethyl nicotinic acid, acmc-20anv9
PubChem CID 28307766
IUPAC Name 2-fluoro-4-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES C1=CN=C(C(=C1C(F)(F)F)C(=O)O)F
Specifications
Melting Point 91°C to 93°C
Quantity 1 g
Formula Weight 209.1
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)nicotinic acid