Learn More
Thermo Scientific Alfa Aesar 50ML 2-(Ethoxycarbonyl)phenylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal^
Brand: Thermo Scientific Alfa Aesar H58721.AD
50ML 2-(Ethoxycarbonyl)phenylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal^
Additional Details : CAS Number : 131379-40-3 Weight : 0.00050kg Transport : UN number : 3399 Chem class : 4.3 Pack group : II