Learn More
Alfa Aesar™ 2-Chloro-4-(trifluoromethyl)pyrimidine, 97%
92.00 EUR - 315.00 EUR
Chemical Identifiers
CAS | 33034-67-2 |
---|---|
Molecular Formula | C5H2ClF3N2 |
Molecular Weight (g/mol) | 182.53 |
MDL Number | MFCD00115111 |
InChI Key | FZRBTBCCMVNZBD-UHFFFAOYSA-N |
Synonym | 2-chloro-4-trifluoromethyl pyrimidine, pyrimidine, 2-chloro-4-trifluoromethyl, 4-trifluoromethyl-2-chloropyrimidine, 2-chloro-4-trifluoromethyl-pyrimidine, pubchem6907, acmc-209hxy, ksc222e4j, abbypharma ap-10-1130 |
PubChem CID | 2773912 |
IUPAC Name | 2-chloro-4-(trifluoromethyl)pyrimidine |
SMILES | C1=CN=C(N=C1C(F)(F)F)Cl |
Description
Chemical Identifiers
33034-67-2 | |
182.53 | |
FZRBTBCCMVNZBD-UHFFFAOYSA-N | |
2773912 | |
C1=CN=C(N=C1C(F)(F)F)Cl |
C5H2ClF3N2 | |
MFCD00115111 | |
2-chloro-4-trifluoromethyl pyrimidine, pyrimidine, 2-chloro-4-trifluoromethyl, 4-trifluoromethyl-2-chloropyrimidine, 2-chloro-4-trifluoromethyl-pyrimidine, pubchem6907, acmc-209hxy, ksc222e4j, abbypharma ap-10-1130 | |
2-chloro-4-(trifluoromethyl)pyrimidine |
Specifications
33034-67-2 | |
49°C (120°F) | |
MFCD00115111 | |
2-chloro-4-trifluoromethyl pyrimidine, pyrimidine, 2-chloro-4-trifluoromethyl, 4-trifluoromethyl-2-chloropyrimidine, 2-chloro-4-trifluoromethyl-pyrimidine, pubchem6907, acmc-209hxy, ksc222e4j, abbypharma ap-10-1130 | |
C1=CN=C(N=C1C(F)(F)F)Cl | |
182.53 | |
182.53 | |
2-Chloro-4-(trifluoromethyl)pyrimidine |
60°C (10 mmHg) | |
C5H2ClF3N2 | |
UN1993 | |
FZRBTBCCMVNZBD-UHFFFAOYSA-N | |
2-chloro-4-(trifluoromethyl)pyrimidine | |
2773912 | |
97% |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.