Learn More
2,4-Dichloronicotinic acid, 97%, Thermo Scientific™
Product Code. 15484558
Click to view available options
Quantity:
250 mg
1 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 262423-77-8
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 192.00
MDL Number MFCD06796292
InChI Key ZFGZNVSNOJPGDV-UHFFFAOYSA-N
Synonym 2,4-dichloronicotinic acid, 2,4-dichloronicotinc acid, 2,4-dichloronicotinicacid, 2,4-dichloro-3-pyridinecarboxylic acid, 2.4-dichloronicotinic acid, 2,4-dichloropyridine-3-carboxylicacid, dichloronicotinic acid;, pubchem17746, 2,4-dichloronicotinic acid;
PubChem CID 10583887
IUPAC Name 2,4-dichloropyridine-3-carboxylic acid
SMILES OC(=O)C1=C(Cl)C=CN=C1Cl
Specifications
Melting Point 166°C to 167°C
Quantity 1 g
Formula Weight 192
Percent Purity 97%
Chemical Name or Material 2,4-Dichloronicotinic acid