missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',4',6'-Trimethylacetophenone, 97+%, Thermo Scientific Chemicals
73.90 EUR - 226.00 EUR
Chemical Identifiers
CAS | 1667-01-2 |
---|---|
Molecular Formula | C11H14O |
Molecular Weight (g/mol) | 162.23 |
MDL Number | MFCD00008735 |
InChI Key | XWCIICLTKWRWCI-UHFFFAOYSA-N |
Synonym | 1-mesitylethanone, 2',4',6'-trimethylacetophenone, acetylmesitylene, acetomesitylene, mesityl methyl ketone, 1-2,4,6-trimethylphenyl ethanone, ethanone, 1-2,4,6-trimethylphenyl, 2-acetylmesitylene, 2,4,6-trimethylacetophenone, methyl 2,4,6-trimethylphenyl ketone |
PubChem CID | 15461 |
IUPAC Name | 1-(2,4,6-trimethylphenyl)ethanone |
SMILES | CC(=O)C1=C(C)C=C(C)C=C1C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11424583
|
Thermo Scientific Alfa Aesar
A11549.14 |
25 g |
73.90 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11434583
|
Thermo Scientific Alfa Aesar
A11549.22 |
100 g |
226.00 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2',4',6'-Trimethylacetophenone is used as laboratory chemicals, manufacture of substances and intermediated for chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2′,4′,6′-Trimethylacetophenone is used as laboratory chemicals, manufacture of substances and intermediated for chemical synthesis.
Solubility
Difficult to mix.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
1667-01-2 | |
162.23 | |
XWCIICLTKWRWCI-UHFFFAOYSA-N | |
15461 | |
CC(=O)C1=C(C)C=C(C)C=C1C |
C11H14O | |
MFCD00008735 | |
1-mesitylethanone, 2',4',6'-trimethylacetophenone, acetylmesitylene, acetomesitylene, mesityl methyl ketone, 1-2,4,6-trimethylphenyl ethanone, ethanone, 1-2,4,6-trimethylphenyl, 2-acetylmesitylene, 2,4,6-trimethylacetophenone, methyl 2,4,6-trimethylphenyl ketone | |
1-(2,4,6-trimethylphenyl)ethanone |
Specifications
1667-01-2 | |
235°C to 236°C | |
C11H14O | |
MFCD00008735 | |
1-mesitylethanone, 2',4',6'-trimethylacetophenone, acetylmesitylene, acetomesitylene, mesityl methyl ketone, 1-2,4,6-trimethylphenyl ethanone, ethanone, 1-2,4,6-trimethylphenyl, 2-acetylmesitylene, 2,4,6-trimethylacetophenone, methyl 2,4,6-trimethylphenyl ketone | |
XWCIICLTKWRWCI-UHFFFAOYSA-N | |
1-(2,4,6-trimethylphenyl)ethanone | |
15461 | |
98% |
0.975 | |
>110°C (230°F) | |
1.517 | |
2044595 | |
Difficult to mix. | |
CC(=O)C1=C(C)C=C(C)C=C1C | |
162.23 | |
162.23 | |
2',4',6'-Trimethylacetophenone |
Safety and Handling
missing translation for 'einecsNumber' : 216-783-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only