Learn More
Pyrex™ Reservoir Assembly
Pyrex™ Reservoir Assembly for use with QWS4 Series Water Still.
Brand: Scilabware WSR20
622.25 EUR valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 6.00000kg
Description
- 20 L
- For use with QWS4 Series Water Stills
Specifications
QWS4 Series Water Still | |
Reservoir Assembly | |
20 L |