Learn More
Reagecon 100ML Osmolality Standard 100mOsm/Kg H2O in 100mlbottle - 1 year shelf-life
Brand:  Reagecon RE-OSM10001
100ML Osmolality Standard 100mOsm/Kg H2O in 100mlbottle - 1 year shelf-life
Product Code. 16421094
  • 257.00 EUR / 100mL