missing translation for 'onlineSavingsMsg'
Learn More

1-Tetradecanethiol, 94%, Thermo Scientific Chemicals

Catalog Number p-7045099
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
2079-95-0
C14H30S
230.454
MFCD00022099
GEKDEMKPCKTKEC-UHFFFAOYSA-N
1-tetradecanethiol, myristyl mercaptan, tetradecanethiol, n-tetradecyl mercaptan, tetradecyl mercaptan, 1-mercaptotetradecane, unii-apy3p7s1xm, apy3p7s1xm, tetradecanethiol-1, acmc-1cq2y
64677
tetradecane-1-thiol
CCCCCCCCCCCCCCS
This item is not returnable. View return policy
2079-95-0
C14H30S
230.454
MFCD00022099
GEKDEMKPCKTKEC-UHFFFAOYSA-N
1-tetradecanethiol, myristyl mercaptan, tetradecanethiol, n-tetradecyl mercaptan, tetradecyl mercaptan, 1-mercaptotetradecane, unii-apy3p7s1xm, apy3p7s1xm, tetradecanethiol-1, acmc-1cq2y
64677
tetradecane-1-thiol
CCCCCCCCCCCCCCS

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 6°C to 7°C
Density 0.85
Boiling Point 176°C to 180°C (22 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Linear Formula CH3(CH2)13SH
Refractive Index 1.461
Quantity 5 g
Beilstein 1742653
Sensitivity Air Sensitive
Formula Weight 230.46
Percent Purity 94%
Chemical Name or Material 1-Tetradecanethiol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'