missing translation for 'onlineSavingsMsg'
Learn More

1-Hexanethiol, 96%, Thermo Scientific Chemicals

Catalog Number p-7045165
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
111-31-9
C6H14S
118.24
MFCD00004909
PMBXCGGQNSVESQ-UHFFFAOYSA-N
1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan
8106
hexane-1-thiol
CCCCCCS
This item is not returnable. View return policy
111-31-9
C6H14S
118.24
MFCD00004909
PMBXCGGQNSVESQ-UHFFFAOYSA-N
1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan
8106
hexane-1-thiol
CCCCCCS

It is used to tune the electrical characteristics in networked carbon nanotube field-effect transistors.1 Also used as a monolayer on copper surfaces to retard surface oxidation during exposure to the environment which can then be desorbed effectively with an annealing step in inert N2 to provide a clean Cu surface.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to tune the electrical characteristics in networked carbon nanotube field-effect transistors.1 Also used as a monolayer on copper surfaces to retard surface oxidation during exposure to the environment which can then be desorbed effectively with an annealing step in inert N2 to provide a clean Cu surface.

Solubility
Difficult to mix in water.

Notes
Air sensitive. Store in cool dry condition. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -81°C
Density 0.838
Boiling Point 150°C to 154°C
Flash Point 30°C (86°F)
Odor Strong
Linear Formula CH3(CH2)5SH
Refractive Index 1.448
Quantity 25 mL
UN Number UN3071
Beilstein 1731295
Sensitivity Air Sensitive
Solubility Information Difficult to mix in water.
Formula Weight 118.24
Percent Purity 97%
Chemical Name or Material 1-Hexanethiol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'