Learn More
(+/-)-1-Boc-2-(aminomethyl)pyrrolidine, tech. 85%, Thermo Scientific™
Product Code. 11442100
Click to view available options
Quantity:
250 mg
1 g
5 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 177911-87-4
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD01317793
InChI Key SOGXYCNKQQJEED-UHFFFAOYSA-N
Synonym tert-butyl 2-aminomethyl pyrrolidine-1-carboxylate, 2-aminomethyl-1-n-boc-pyrrolidine, +/--2-aminomethyl-1-n-boc-pyrrolidine, 1-boc-2-aminomethyl-pyrrolidine, 1-boc-2-aminomethyl pyrrolidine, 1-n-boc-2-aminomethyl pyrrolidine, 2-aminomethyl-pyrrolidine-1-carboxylic acid tert-butyl ester, 1-boc-aminomethyl pyrrolidine, 1-boc-2-aminomethylpyrrolidine
PubChem CID 2756484
IUPAC Name tert-butyl 2-(aminomethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1CN
Specifications
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 200.28
Percent Purity 85%
Grade Technical
Chemical Name or Material (+/-)-1-Boc-2-(aminomethyl)pyrrolidine