Learn More
1-(6-Trifluoromethyl-2-pyridyl)-1,4-diazepane, 95%, Thermo Scientific™
89.50 EUR - 251.00 EUR
Chemical Identifiers
CAS | 243666-15-1 |
---|---|
Molecular Formula | C11H14F3N3 |
Molecular Weight (g/mol) | 245.249 |
MDL Number | MFCD12756231 |
InChI Key | BMKFDQXBPCCAFY-UHFFFAOYSA-N |
Synonym | 1-3-trifluoromethyl pyridin-2-yl-1,4-diazepane, 1-3-trifluoromethyl pyrid-2-yl-1,4-diazepane, 1-3-trifluoromethyl pyridin-2-yl homopiperazine, 1-3-trifluoromethyl-2-pyridyl-1,4-diazaperhydroepine |
PubChem CID | 2778749 |
IUPAC Name | 1-[3-(trifluoromethyl)pyridin-2-yl]-1,4-diazepane |
SMILES | C1CNCCN(C1)C2=C(C=CC=N2)C(F)(F)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
243666-15-1 | |
245.249 | |
BMKFDQXBPCCAFY-UHFFFAOYSA-N | |
2778749 | |
C1CNCCN(C1)C2=C(C=CC=N2)C(F)(F)F |
C11H14F3N3 | |
MFCD12756231 | |
1-3-trifluoromethyl pyridin-2-yl-1,4-diazepane, 1-3-trifluoromethyl pyrid-2-yl-1,4-diazepane, 1-3-trifluoromethyl pyridin-2-yl homopiperazine, 1-3-trifluoromethyl-2-pyridyl-1,4-diazaperhydroepine | |
1-[3-(trifluoromethyl)pyridin-2-yl]-1,4-diazepane |
Specifications
243666-15-1 | |
1.521 | |
UN3267 | |
BMKFDQXBPCCAFY-UHFFFAOYSA-N | |
1-[3-(trifluoromethyl)pyridin-2-yl]-1,4-diazepane | |
2778749 | |
95% |
C11H14F3N3 | |
MFCD12756231 | |
1-3-trifluoromethyl pyridin-2-yl-1,4-diazepane, 1-3-trifluoromethyl pyrid-2-yl-1,4-diazepane, 1-3-trifluoromethyl pyridin-2-yl homopiperazine, 1-3-trifluoromethyl-2-pyridyl-1,4-diazaperhydroepine | |
C1CNCCN(C1)C2=C(C=CC=N2)C(F)(F)F | |
245.249 | |
245.25 | |
1-(6-Trifluoromethyl-2-pyridyl)-1,4-diazepane |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures