Learn More
1,3-Dichlorobenzene, 98%, Thermo Scientific Chemicals
49.80 EUR - 129.00 EUR
Chemical Identifiers
CAS | 541-73-1 |
---|---|
Molecular Formula | C6H4Cl2 |
Molecular Weight (g/mol) | 146.998 |
MDL Number | MFCD00000573 |
InChI Key | ZPQOPVIELGIULI-UHFFFAOYSA-N |
Synonym | m-dichlorobenzene, m-phenylene dichloride, m-dichlorobenzol, meta-dichlorobenzene, benzene, 1,3-dichloro, metadichlorobenzene, benzene, m-dichloro, m-phenylenedichloride, m-dcb, 3-dichlorobenzene |
PubChem CID | 10943 |
ChEBI | CHEBI:36693 |
IUPAC Name | 1,3-dichlorobenzene |
SMILES | C1=CC(=CC(=C1)Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11434664
|
Thermo Scientific Alfa Aesar
A13688.30 |
250 g |
49.80 EUR
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11454230
|
Thermo Scientific Alfa Aesar
A13688.36 |
500 g |
82.90 EUR
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11424664
|
Thermo Scientific Alfa Aesar
A13688.0B |
1000 g | |||||||
Description
1,3-Dichlorobenzene is used in medicine, and dyes. And it is also used to study the effect of solvent vapor pressure on the vertical nanowire of C60 molecules1. 1,3-Dichlorobenzene, a chlorinated aromatic hydrocarbon, can be detected in environmental samples using an advanced Fourier transform infrared spectroscopy-attenuated total reflectance (FTIR-ATR) sensor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3-Dichlorobenzene is used in medicine, and dyes. And it is also used to study the effect of solvent vapor pressure on the vertical nanowire of C60 molecules1. 1,3-Dichlorobenzene, a chlorinated aromatic hydrocarbon, can be detected in environmental samples using an advanced Fourier transform infrared spectroscopy-attenuated total reflectance (FTIR-ATR) sensor.
Solubility
Difficult to mix.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
541-73-1 | |
146.998 | |
ZPQOPVIELGIULI-UHFFFAOYSA-N | |
10943 | |
1,3-dichlorobenzene |
C6H4Cl2 | |
MFCD00000573 | |
m-dichlorobenzene, m-phenylene dichloride, m-dichlorobenzol, meta-dichlorobenzene, benzene, 1,3-dichloro, metadichlorobenzene, benzene, m-dichloro, m-phenylenedichloride, m-dcb, 3-dichlorobenzene | |
CHEBI:36693 | |
C1=CC(=CC(=C1)Cl)Cl |
Specifications
541-73-1 | |
1.288 | |
63°C (145°F) | |
1.546 | |
UN3082 | |
14,3055 | |
Difficult to mix. | |
C1=CC(=CC(=C1)Cl)Cl | |
146.998 | |
CHEBI:36693 | |
98% |
-25°C | |
173°C | |
C6H4Cl2 | |
MFCD00000573 | |
956618 | |
m-dichlorobenzene, m-phenylene dichloride, m-dichlorobenzol, meta-dichlorobenzene, benzene, 1,3-dichloro, metadichlorobenzene, benzene, m-dichloro, m-phenylenedichloride, m-dcb, 3-dichlorobenzene | |
ZPQOPVIELGIULI-UHFFFAOYSA-N | |
1,3-dichlorobenzene | |
10943 | |
147 | |
1,3-Dichlorobenzene |
Safety and Handling
GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
P210-P233-P240-P243-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P374-P501c
H227-H302-H332
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
missing translation for 'einecsNumber' : 208-792-1
missing translation for 'rtecsNumber' : CZ4499000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only