missing translation for 'onlineSavingsMsg'
Learn More

[1,3-Bis(diphenylphosphino)propane]nickel(II) chloride, 98+%, Thermo Scientific Chemicals

Catalog Number p-3735620
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
15629-92-2
C27H26Cl2NiP2
542.04
MFCD00015318
REQVOKPBMSCTNI-UHFFFAOYSA-L
131675641
3-diphenylphosphanylpropyl(diphenyl)phosphane;nickel;dihydrochloride
Cl[Ni++]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
15629-92-2
C27H26Cl2NiP2
542.04
MFCD00015318
REQVOKPBMSCTNI-UHFFFAOYSA-L
131675641
3-diphenylphosphanylpropyl(diphenyl)phosphane;nickel;dihydrochloride
Cl[Ni++]Cl.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 213.0°C
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Fieser 06,50; 15,122
Solubility Information Solubility in water: insoluble
Formula Weight 542.04
Percent Purity 99%
Physical Form Crystals or Powder
Chemical Name or Material [1,3-Bis(diphenylphosphino)propane]nickel(II) chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • ¢1,3-Bis(diphenylphosphino)propane!nickel(II) chloride
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H350-May cause cancer.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'